Order Entry
Denmark
Orders LinkContactUsLinkComponent
Plexiglas rod
Plexiglas rod
Plexiglas rod
Catalog # 765-0184
Marketsource
Plexiglas rod
Catalog # 765-0184
Marketsource
Plexiglas rod
Plexiglas rod
Plexiglas rod
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
Used in electrostatic experiments. Equipped with holes, 4 mm, at one end for connection of conductor ball or capacitor plate.
  • Dimensions: 150 × 15 mm
Specifications
  • Description:
    Plexiglas rod