Order Entry
Denmark
Orders LinkContactUsLinkComponent
Modular diode laser
Modular diode laser
Supplier:   KVANT LIFE SCIENCE
Modular diode laser
Supplier:  KVANT LIFE SCIENCE
Certificates
Modular diode laser
Modular diode laser
1 Options Available Below
Product Details & Documents
The advantages of a diode laser compared to a HeNe laser are mainly small format, low power consumption and long service life, as well as the possibility of electronic modulation. The laser is modular in the range 100 Hz 20 MHz. Dimensions.
  • Elastic in width and thus easy to apply
Product Family Options
Product Information
  • Availability
    Price
  • 765-1004
    Modular diode laser, 635 nm, 3 mW
    Modular diode laser, 635 nm, 3 mW
    Discontinued
  • 765-1018
    Modular diode laser, 635 nm, 1 mW
    Modular diode laser, 635 nm, 1 mW
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each