Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Propylene oxide, Supelco®

Flammable
Health
Irritant
Supplier:   Merck
Propylene oxide, Supelco®
Supplier:  Merck
Certificates
Propylene oxide, Supelco®
Propylene oxide, Supelco®

2 Options Available Below

Product Details & Documents
Synonyms:  (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
  • CAS Number:75-56-9
  • Formula:C₃H₆O
  • MDL Number:MFCD00005126
  • Molecular Weight:58.08 g/mol
  • Merck Index:12,08041
  • Boiling Pt:34.3 °C (1013 hPa)
  • Melting Pt:–112 °C
  • Density:0.83 g/cm³ (20 °C)
  • Flash Pt:–37 °C
  • Storage Temperature:Refrigerator
  • UN:1280
  • ADR:3,I
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-879-2

 

Product Family Options

Product Information