Order Entry
Denmark
ContactUsLinkComponent
Sodium carbonate 0.5 M Na2CO3 (1N) volumetric solution, Fluka™
Sodium carbonate 0.5 M Na2CO3 (1N) volumetric solution, Fluka™
Catalog # 34277-1L
CAS Number:  
undefined
Sodium carbonate 0.5 M Na2CO3 (1N) volumetric solution, Fluka™
Catalog # 34277-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

Synonyms: Calcined soda, Carbonic acid disodium salt
 :
 :105.99 g/mol
Storage Temperature:Ambient
EINECS:415-456-6
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494