Order Entry
Denmark
ContactUsLinkComponent
Bovine thrombin (from plasma)
Bovine thrombin (from plasma)
Supplier:  Serva
Certificates
Bovine thrombin (from plasma)
Supplier:  Serva
Certificates

Specifications

  • CAS No.:
    9002-04-4
  • Source:
    Plasma
  • Species:
    Bovine
  • Enzyme name:
    Thrombin

Specifications

Related Information

Frequently Bought Together