Pepsin
Glass bottle for solids, 100 g
Catalog # SERA31825.01
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Cat. No.:SERA31825.01
- Storage conditions:Refrigerator
- Enzyme name:Pepsin
- CAS No.:9001-75-6
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



