Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Aniline 1000 µg/ml in methanol
Organic reference standard, Aniline 1000 µg/ml in methanol
Catalog # REST31470-3
Supplier:  Restek
CAS Number:  
Organic reference standard, Aniline 1000 µg/ml in methanol
Catalog # REST31470-3
Supplier:  Restek
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    1 Ampoul
  • CAS No.:
    62-53-3
  • Description:
    Aniline 1,000 µg/ml in methanol
  • Formula:
    C₆H₇N
  • Description:
    Aniline

Specifications

About this item