Order Entry
Denmark
ContactUsLinkComponent
Pesticide reference standard, Pentachlorophenol 1000 µg/ml in methanol
Pesticide reference standard, Pentachlorophenol 1000 µg/ml in methanol
Supplier:  Restek
Certificates
Pesticide reference standard, Pentachlorophenol 1000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    87-86-5
  • Formula:
    C₆Cl₅OH 
  • Description:
    Pentachlorophenol

Specifications

Frequently Bought Together