Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, Isobutanol 2,000 µg/ml in methanol
 :  Restek
 
Organic reference standard, Isobutanol 2,000 µg/ml in methanol
 :  Restek

 

  • CAS No.:
    78-83-1
  • Formula:
    C₄H₁₀O
  • Description:
    Isobutanol

 

Frequently Bought Together