3,3',5,5'-Tetramethylbenzidine (TMB) ≥99%, GR for analysis, Supelco®
1.08622.0001
: Merck
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Assay (perchloric acid titration):≥ 99 %
- Identity (IR-spectrum):conforms
- Melting range:166 - 170 °C
- Loss on Drying (105°C):≤ 0.2 %
- Cd (Cadmium):Max. 0.15 ppm
:
:240.35 g/mol
:402 °C (1013 hPa)
:166…170 °C
:M-Clarity™ quality level = MQ100
:Ambient
:259-364-6
:54827-17-7
:C₁₆H₂₀N₂
:MFCD00007748