Order Entry
Denmark
ContactUsLinkComponent
Nitric acid 0.1 M HNO3 (0.1N) volumetric solution, Fluka™
Corrosive
Nitric acid 0.1 M HNO3 (0.1N) volumetric solution, Fluka™
Catalog # 35318-1L
CAS Number:  
Corrosive
Nitric acid 0.1 M HNO3 (0.1N) volumetric solution, Fluka™
Catalog # 35318-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
EINECS:231-791-2
UN:3264
ADR:8,III
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349