3,3',5,5'-Tetramethylbenzidine (TMB), certified
Catalog # EHERC17413000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:240.35 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:166…170 °C
Storage Temperature:Ambient
EINECS:259-364-6
CAS Number:54827-17-7
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748