Ammonium acetate ≥97.0%, crystals, AR® ACS, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
:77.08 g/mol
Merck Index:13,00495
Melting Pt:114 °C
Density:1.17 g/cm³ (20 °C)
Flash Pt:136 °C
EINECS:211-162-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:≥97%
- ACS - Assay (CH₃COONH₄):≥97 %
- ACS - Appearance (colorless crystals):Passes Test
- ACS - Chloride (Cl):≤5 ppm
- ACS - Heavy Metals (as Pb):≤5 ppm
- ACS - Insoluble Matter:≤0.005 %
- ACS - Iron (Fe):≤5 ppm
- ACS - Nitrate (NO₃):≤0.001 %
- ACS - pH of 5% Solution at 25°C:6.7 - 7.3
- ACS - Residue after Ignition:≤0.01 %
- ACS - Sulfate (SO₄):≤0.001 %
Specification Test Results
Frequently Bought Together