Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Safranine O
Safranine O for microscopy (C.I. 50240)
Safranine O for microscopy (C.I. 50240)
Safranine O for microscopy (C.I. 50240)
APPCA0427.0100
:
PanReac AppliChem
:
Safranine O for microscopy (C.I. 50240)
APPCA0427.0100
:
PanReac AppliChem
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
350.85 g/mol
:
1.447 g/cm³ (22 °C)
:
Ambient
:
207-518-8
:
477-73-6
:
C₂₀H₁₉ClN₄
:
MFCD00011759