Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

D-(-)-Ribose ≥98.0%, high purity

100 g
Catalog # 0671-100G
Supplier:   VWR Chemicals
D-(-)-Ribose ≥98.0%, high purity
Catalog # 0671-100G
Supplier:  VWR Chemicals
CAS Number:  50-69-1
D-(-)-Ribose ≥98.0%, high purity
D-(-)-Ribose ≥98.0%, high purity

100 g

CAS Number:  50-69-1
Price
926,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:50-69-1
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00135453
  • Molecular Weight:150.13 g/mol
  • Merck Index:12,08371
  • Boiling Pt:331 °C (1013 hPa)
  • Melting Pt:90…95 °C
  • Density:0.8 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-059-4
Specifications

Specification Test Results

  • Arsenic:
    <0.0002%
  • Ash:
    ≤0.2%
  • Heavy Metals (as Pb):
    <0.001%
  • Loss on Drying:
    ≤3.0%
  • Purity:
    >98.0%
  • Specific Rotation (4%, Water):
    (–21.5) to (–19.7)°

Specifications

  • Pk:
    100 g

Specifications