(±)-2-Methyltetrahydrofuran ≥99% stabilised
Leverandør: Thermo Fisher Scientific
Gå til:
Om denne vare
Stabilized with ca 150-400 ppm BHT
Purity: 99% CAS No.: 96-47-9 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: -11°(12°F) Refractive Index: 1.4060 Melting Point: -136° Boiling Point: 80-82° Beilstein Reference: 102448 UN No.: 2536 MDL No.: MFCD00005367
- CAS nummer:96-47-9
- Formel:C₅H₁₀O
- MDL nummer:MFCD00005367
- Molecular Weight:86.13 g/mol
- Kogende Pt:80…82 °C
- Smeltende Pt:–136 °C
- Tæthed:0.856
- Flash Pt:–11 °C (–12 °F)
- Opbevaringstemperatur:Ambient
- EINECS:202-507-4
- ADR:3,II
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 150-400 ppm
- FN:2536
2 tilgængelige muligheder nedenfor
Product Details & Documents
Stabilized with ca 150-400 ppm BHT
Purity: 99% CAS No.: 96-47-9 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: -11°(12°F) Refractive Index: 1.4060 Melting Point: -136° Boiling Point: 80-82° Beilstein Reference: 102448 UN No.: 2536 MDL No.: MFCD00005367
- CAS nummer:96-47-9
- Formel:C₅H₁₀O
- MDL nummer:MFCD00005367
- Molecular Weight:86.13 g/mol
- Kogende Pt:80…82 °C
- Smeltende Pt:–136 °C
- Tæthed:0.856
- Flash Pt:–11 °C (–12 °F)
- Opbevaringstemperatur:Ambient
- EINECS:202-507-4
- ADR:3,II
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 150-400 ppm
- FN:2536



