
1H,1H,2H,2H-Perfluoro-1-iodohexane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2043-55-2
:
Formula:C₆H₄F₉I
MDL Number:MFCD00039409
:373.99 g/mol
Boiling Pt:138…140 °C
Melting Pt:–25 °C
Density:1.940
Flash Pt:>110 °C (230 °F)
EINECS:218-055-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together