Squalene ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 111-02-4 Molecular Formula: C30H50 Formula Weight: 410.73 Storage Temperature: 2°C to 8°C Flash Point: 218°(424°F) Refractive Index: 1.4955 Melting Point: -75° Boiling Point: 285°/25mm Merck Reference: 14,8768 MDL No.: MFCD00008912
Biosynthetic precursor to all steroids
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
- CAS Number:111-02-4
- Formula:C₃₀H₅₀
- MDL Number:MFCD00008912
- Molecular Weight:410.73 g/mol
- Merck Index:14,8768
- Boiling Pt:285 °C (25 mmHg)
- Melting Pt:–75 °C
- Density:0.855
- Flash Pt:218 °C (424 °F)
- EINECS:203-826-1
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 111-02-4 Molecular Formula: C30H50 Formula Weight: 410.73 Storage Temperature: 2°C to 8°C Flash Point: 218°(424°F) Refractive Index: 1.4955 Melting Point: -75° Boiling Point: 285°/25mm Merck Reference: 14,8768 MDL No.: MFCD00008912
Biosynthetic precursor to all steroids
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
- CAS Number:111-02-4
- Formula:C₃₀H₅₀
- MDL Number:MFCD00008912
- Molecular Weight:410.73 g/mol
- Merck Index:14,8768
- Boiling Pt:285 °C (25 mmHg)
- Melting Pt:–75 °C
- Density:0.855
- Flash Pt:218 °C (424 °F)
- EINECS:203-826-1



