Diethylene glycol monobutyl ether ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-(2-Butoxyethoxy)ethanol, Diethylene glycol mono-n-butyl ether, Butyldigol
CAS Number:112-34-5
:
Formula:CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MDL Number:MFCD00002881
:162.23 g/mol
Merck Index:14,1557
Boiling Pt:230…232 °C
Melting Pt:–68 °C
Density:0.967
Flash Pt:100 °C (212 °F)
EINECS:203-961-6
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together