Order Entry
Denmark
ContactUsLinkComponent
Silver hexafluoroarsenate ≥98%
Toxic
Environment
Silver hexafluoroarsenate ≥98%
Catalog # 11608.09
CAS Number:  
Toxic
Environment
Silver hexafluoroarsenate ≥98%
Catalog # 11608.09
CAS Number:  

Specifications

  • Pk:
    10 g

Specifications

About this item

 :
 :296.78
Storage Temperature:Ambient
UN:1557
ADR:6.1,II
 :GHS06
 :GHS09
CAS Number:12005-82-2
Formula:AgAsF₆
MDL Number:MFCD00014142