
1-Hexadecyl-3-methylimidazolium chloride monohydrate 98%
Catalog # ACRO356650100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Light yellow Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %
- Water:3 to 5 % (K.F.)
Specification Test Results
About this item
:
:361.01 g/mol
:GHS07
CAS Number:404001-62-3
Formula:C₂₀H₃₉ClN₂·1H₂O
MDL Number:MFCD07773044