Order Entry
Denmark
ContactUsLinkComponent
2-Amino-2-methyl-1,3-propanediol 99%
Irritant
Corrosive
2-Amino-2-methyl-1,3-propanediol 99%
Certificates
undefined
Irritant
Corrosive
2-Amino-2-methyl-1,3-propanediol 99%

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    ≥100 °C
  • Water:
    ≤0.5 %
  • Titration with HClO4:
    ≥98.5 % (on anhydrous substance)

Specification Test Results

Related Information

Frequently Bought Together