Order Entry
Denmark
Orders LinkContactUsLinkComponent
Demonstration range for oh
  765-0116
Marketsource
Demonstration range for oh
  765-0116
Marketsource
Demonstration range for oh
Demonstration range for oh
1 * 1 Each
Demonstration model of a caliper. Made of transparent plastic. Equipped with Nonies scale and rubber feet. Intended for OHapparat. Can also be used independently.
  • Dimensions: 200 × 105 mm