Order Entry
Denmark
Orders LinkContactUsLinkComponent
Storage systems
Storage systems
Supplier:   GRATNELL'S
Storage systems
Supplier:  GRATNELL'S
Certificates
Storage systems
3 Options Available Below
Product Details & Documents
Instead of cabinets and shelving systems. Very practical to store all laboratory and demonstration equipment in. When used, just take out a tray or a shelf and load directly into the trolley (see art. no. 765-0935 for example). Comes with roof section and adjustable feet.
Slopes and hill holders will be added.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 765-0937
    Storage rack, 2 sections
    Storage rack, 2 sections
    710×420×1850 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.780,00 kr
    9.780,00 kr
    1 * 1 Items
       9.780,00 kr
  • 765-0939
    Storage rack, 3 sections, low
    Storage rack, 3 sections, low
    1055×420×1500 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5.440,00 kr
    5.440,00 kr
    1 * 1 Items
       5.440,00 kr
  • 765-0941
    Storage rack, 3 sections
    Storage rack, 3 sections
    1055×420×1850 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 12.090,00 kr
    12.090,00 kr
    1 * 1 Items
       12.090,00 kr
  • 765-0940
    Shelves for storage racks
    Shelves for storage racks
    Discontinued