Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

trans-4-Hydroxy-L(-)-proline

Glass bottle for solids, 5 g
Catalog # ROTH3893.1
Marketsource
Supplier:   Roth Carl
trans-4-Hydroxy-L(-)-proline
Catalog # ROTH3893.1
Supplier:  Roth Carl
Marketsource
CAS Number:  51-35-4
trans-4-Hydroxy-L(-)-proline

Glass bottle for solids, 5 g

CAS Number:  51-35-4
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
  • CAS Number:51-35-4
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064320
  • Molecular Weight:131.13 g/mol
  • Merck Index:13,04866
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:206 °C
  • Storage Temperature:Ambient
  • EINECS:200-091-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications