Order Entry
Denmark
ContactUsLinkComponent
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride 95%
Corrosive
Irritant
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride 95%
Catalog # APOSOR315373-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride 95%
Catalog # APOSOR315373-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :222.11 g/mol
Melting Pt:128 - 131 °C
 :GHS05
 :GHS07
CAS Number:86604-75-3
Formula:C₉H₁₂ClNO
MDL Number:MFCD00277472
Melting Pt:128…131 °C