Order Entry
Denmark
ContactUsLinkComponent
VWR® Sodium Acetate Buffer 3,0 M Solution, pH 5,2
VWR® Sodium Acetate Buffer 3,0 M Solution, pH 5,2
  E521-100ML
 :  
undefined
VWR® Sodium Acetate Buffer 3,0 M Solution, pH 5,2
  E521-100ML
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Sodium acetate buffer solution pH 5.2 (3 mol/l)
With diethyl pyrocarbonate (DEPC).
  • DNase-free
  • RNase-free