
6-TET phosphoramidite [5'-Tetrachlorofluorescein phosphoramidite] ≥95%
Supplier: AAT BIOQUEST
Certificates
About this item
CAS Number:877049-90-6
:
Formula:C₄₆H₅₄N₃O₁₀Cl₄P
MDL Number:MFCD01940926
:981.72 g/mol
ADR:5,II
:GHS07
Frequently Bought Together