0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Phalloidin-PEG4-DBCO
Phalloidin-PEG4-DBCO for biomolecular labeling
Phalloidin-PEG4-DBCO for biomolecular labeling
AATB5303
:
AAT BIOQUEST
:
Phalloidin-PEG4-DBCO for biomolecular labeling
AATB5303
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
100 µG
Seq:
1286
:
:
5,II
:
GHS07