Spring til Overskrift
Spring til Hovedindhold
Spring til Sidefod
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AF532 PEG4 biotin conjugate
AF532 PEG4 biotin conjugate
AF532 PEG4 biotin conjugate
Katalog # AATB3110
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
AF532 PEG4 biotin conjugate
Katalog # AATB3110
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
1172.48 g/mol
Product Details