Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Log ind
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Preactivated APC
Preactivated APC, ReadiUse™ fluorescent probe, preactivated
Preactivated APC, ReadiUse™ fluorescent probe, preactivated
Irritant
Preactivated APC, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2561
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
Preactivated APC, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2561
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details