Order Entry
Denmark
ContactUsLinkComponent
 
Irritant
BCECF AM ester calcium indicator
Catalog # AATB21202
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
BCECF AM ester calcium indicator
Catalog # AATB21202
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :808.69 g/mol
ADR:5,II
 :GHS07
CAS Number:117464-70-7
Formula:C₄₂H₄₀O₂₁
MDL Number:MFCD00036969