Cyanine 5 azide ≥90% [equivalent to Cy5® azide]
Supplier: AAT BIOQUEST
Certificates
About this item
:
:838.91 g/mol
Frequently Bought Together
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
Thermo Scientific™ Matrix™ Sample Storage Starter Packs
Supplier: THERMO MATRIX TECHNOLOGIES
Micro packaging vials with thread, Nalgene®
Supplier: Thermo Fisher Scientific
HTS TranswellRegistered Mark-96, 0.4 µm Pore Polyester Membrane Inserts 1 * 480 items
Supplier: STEMCELL Technologies
Packaging and Storage Boxes
Supplier: SEMADENI
Detectable scoops, SteriPlast®
Supplier: BURKLE
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning
Filter plates, 96-well, AcroPrep™
Supplier: Cytiva (Formerly Pall Lab)
Pasteur Pipette,Extended Fine Tip,3.3ml Sterile 1 * 500 items
Supplier: DELTALAB
Seals for ALPS™ 50 V heat sealer
Supplier: THERMO MATRIX TECHNOLOGIES