Annexin V, XFD594 Conjugate
AATB20096
:
- Pakn:100 Tests
- Beskrivelse:Annexin V, XFD594 conjugate
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
- High stability
- In-house quality control
- Multiple research applications
: For research use only.