Order Entry
Denmark
ContactUsLinkComponent
Texas Red® Alkyne, Single Isomer
Texas Red® Alkyne, Single Isomer
Katalog # AATB485
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Texas Red® Alkyne, Single Isomer
Katalog # AATB485
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Description:
    Texas Red® alkyne
  • Purity:
    90%

Specifications

About this item

Texas Red® alkyne is an excellent azide-reactive fluorescent reagent for labelling alkyne-containing biological molecules through the well known click chemistry.

  • Identical spectra to Texas Red
  • High Stability
Caution: For research use only.