Order Entry
Denmark
ContactUsLinkComponent
CytoTrace™ Ultra Green
CytoTrace™ Ultra Green
Catalog # AATB21800
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
CytoTrace™ Ultra Green
Catalog # AATB21800
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Description:
    CytoTrace™ ultra green
  • Purity:
    90%

Specifications

About this item

CMFDA (5-chloromethylfluorescein diacetate) has been used as a prominent cell tracker in the research community for a long time.

  • Bright Signal
  • High Sensitivity
  • Fixable
  • Stable Signal
Caution: For research use only.