Cell Explorer™ Live Cell Tracking Kit *Red Fluorescence*
Catalog # AATB22623
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Cell Explorer™ live cell tracking kit *red fluorescence*
Specifications
About this item
The Cell Explorer™ fluorescence imaging kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
- Long label retention
- Suitable for all major fluorescence platforms
- Contains all necessary components
- Multiple applications
Caution: For research use only.