Order Entry
Denmark
ContactUsLinkComponent
Live or Dead™ Fixable Dead Cell Staining Kit *Orange Fluorescence*
Live or Dead™ Fixable Dead Cell Staining Kit *Orange Fluorescence*
Catalog # AATB22602
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Live or Dead™ Fixable Dead Cell Staining Kit *Orange Fluorescence*
Catalog # AATB22602
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Live or Dead™ fixable dead cell staining kit *orange fluorescence*

Specifications

About this item

Our Live or Dead™ Fixable Dead Cell Staining Kits are a set of tools for labelling cells for fluorescence microscopic investigations of cell functions.

  • Contains all necessary components
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.