Live or Dead™ Fixable Dead Cell Staining Kit *Orange Fluorescence with 405 nm Excitation*
Catalog # AATB22502
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Live or Dead™ fixable dead cell staining kit *orange fluorescence with 405 nm excitation*
Specifications
About this item
Our Live or Dead™ fixable dead cell staining kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
- Contains all necessary components
- Bright and stable signal
- Suitable for all major fluorescence platforms
- Large stokes shift
Caution: For research use only.