Order Entry
Denmark
ContactUsLinkComponent
 
Cell Meter™ Multiplexing Caspase 3/7, 8 and 9 Activity Assay Kit *Triple Fluorescence Colors*
Catalog # AATB22820
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ Multiplexing Caspase 3/7, 8 and 9 Activity Assay Kit *Triple Fluorescence Colors*
Catalog # AATB22820
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ multiplexing caspase 3/7, 8 and 9 activity assay kit *triple fluorescence colors*

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.