Screen Quest™ Rhod-4 No Wash Calcium Assay Kits, Medium Removal
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
- >250 times fluorescence increase once bound to calcium
- Long-wavelength signal emission wavelength
- Contains all necessary components
- No wash step required
- Robust and adaptable protocol
- brighter and more photostable signal
: For research use only.
Frequently Bought Together