Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Red Fluorescence Optimized for Microplate Reader*

Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Red Fluorescence Optimized for Microplate Reader*

Cell Meter™ mitochondrion membrane potential assay kit *red fluorescence optimized for microplate reader*, 500 Tests
Catalog # AATB22807
Supplier:   AAT Bioquest
Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Red Fluorescence Optimized for Microplate Reader*
Catalog # AATB22807
Supplier:  AAT Bioquest
CAS Number:  
Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Red Fluorescence Optimized for Microplate Reader*
Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Red Fluorescence Optimized for Microplate Reader*

Cell Meter™ mitochondrion membrane potential assay kit *red fluorescence optimized for microplate reader*, 500 Tests

CAS Number:  
Price
2.350,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimised for microplate readers but suitable for all major fluorescence platforms

Caution: For research use only.

Specifications

Specifications

  • Pk:
    500 Tests
  • Description:
    Cell Meter™ mitochondrion membrane potential assay kit *red fluorescence optimized for microplate reader*

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.