MycoLight™ Live Bacteria Fluorescence Imaging Kit
Catalog # AATB22409
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:100 Tests
- Description:MycoLight™ live bacteria fluorescence imaging kit
- Environmentally Preferable:
Specifications
About this item
The MycoLight™ Live Bacteria Fluorescence Imaging Kit provides an easy and convenient way for visualizing live bacteria through fluorescent microscope.
- Suitable for gram-positive and gram-negative bacteria
- Contains all necessary components
- Multiple applications
- Optimised for fluorescent microscopy
Caution: For research use only.