Cell Meter™ Intracellular GSH Assay Kit *Optimised for Flow Cytometry*
Catalog # AATB22810
Leverandør: AAT BIOQUEST
CAS nummer:
Specifikationer
- Pk:100 Tests
- Description:Cell Meter™ intracellular GSH assay kit *optimized for flow cytometry*
Specifikationer
Om denne vare
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimised for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.