Order Entry
Denmark
ContactUsLinkComponent
Chloroform-[D₁] (99.8% D) +0.03% V/V TMS for NMR spectroscopy
 
 
Chloroform-[D₁] (99.8% D) +0.03% V/V TMS for NMR spectroscopy
  CAMBDLM-7TC-100
 :  
 
 
Chloroform-[D₁] (99.8% D) +0.03% V/V TMS for NMR spectroscopy
  CAMBDLM-7TC-100
 :  

 

  • Pk:
    100 g
  • Seq:
    3401

 

 

 :
 :120.38 g/mol
 :60.8 °C (1013 hPa)
 :–64.1 °C
 :1.5 g/cm³ (20 °C)
 :Refrigerator
 :212-742-4
 :1888
 :6.1,III
 :GHS08
 :GHS07
 :865-49-6
 :CDCl₃
 :MFCD00000827