Gadolinium ≥99% (REO, rare earth oxide basis), powder -40 mesh
:7440-54-2
:
:Gd
:MFCD00011022
:157.25 g/mol
:12,04346
:3273 °C (1013 hPa)
:1312 °C
:7.886 g/cm³ (20 °C)
:3178
:4.1,II
:Packaged under argon.
:231-162-2
:GHS02
:Ambient
Frequently Bought Together