Recombinant Proteinase K
GHS symbols: GHS07; GHS08
GHS signal word: Danger
EINECS: 254-457-8
- CAS No.:39450-01-6
- Enzyme type:Recombinant
- Storage conditions:Freezer (2...8 °C)
- Enzyme name:Proteinase K
- Enzyme synonyms:Recombinant Proteinase K, Recombinant Protease K, Recombinant Endopeptidase K, Recombinant Tritirachium alkaline proteinase, Recombinant Tritirachium album serine proteinase
- Molecular weight:28,8 kD
Frequently Bought Together