Screw Cap Tubes
Supplier: MEGRO
Jump to:
About this item
For secure transport and storage of samples.
- Made of selected, biologically inert polypropylene
- Reliable sealing
- Polyethylene screw cap
- Not autoclavable
2 Options Available Below
Product Details & Documents
For secure transport and storage of samples.
- Made of selected, biologically inert polypropylene
- Reliable sealing
- Polyethylene screw cap
- Not autoclavable
Product Family Options
Product Information
- CapacityColourLengthTube ext.ØCap includedCap colourAvailabilityPrice
- MEGR122120Screwcap tubes with enclosed capScrewcap tubes with enclosed cap13 mlNatural100 mm16 mmYesNaturalLog in for availability1 * 1.000 Items804,00 kr
- MEGR122121Screwcap tubes with fitted capScrewcap tubes with fitted cap13 mlNatural100 mm16 mmYesNaturalLog in for availability1 * 1.000 Items1.070,00 kr
Anbefalinger