Order Entry
Denmark
ContactUsLinkComponent
MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit
MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit
Catalog # AATB22415
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit
Catalog # AATB22415
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    MycoLight™ Rapid fluorescence gram-positive bacteria staining kit
  • Environmentally Preferable:
  • Tests per kit:
    100 Tests

Specifications

About this item

MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit provides a novel one-step fluorescence assay for the determination of gram sign in living bacteria.

  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimized for fluorescent microscopy
Caution: For research use only.