Order Entry
Denmark
ContactUsLinkComponent
MycoLight™ Flow Cytometric Live Bacteria Assay Kit
MycoLight™ Flow Cytometric Live Bacteria Assay Kit
Catalog # AATB22407
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
MycoLight™ Flow Cytometric Live Bacteria Assay Kit
Catalog # AATB22407
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    MycoLight™ flow cytometric live bacteria assay kit
  • Tests per kit:
    100 Tests

Specifications

About this item

The MycoLight Flow cytometric live bacteria assay kit provides an easy and convenient method for evaluating bacterial vitality as a function of the intracellular esterase activity.

  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimized for flow cytometry
Caution: For research use only.